Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W479455

Sigma-Aldrich

1-Cyclohexylethanol

≥95%

Synonym(s):

α-Methylcyclohexanemethanol, (1-Hydroxyethyl)cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H11)OH
CAS Number:
Molecular Weight:
128.21
FEMA Number:
4794
Beilstein/REAXYS Number:
2322053
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

assay

≥95%

refractive index

n20/D 1.465 (lit.)

bp

189 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

clean; fresh

SMILES string

CC(O)C1CCCCC1

InChI

1S/C8H16O/c1-7(9)8-5-3-2-4-6-8/h7-9H,2-6H2,1H3

InChI key

JMSUNAQVHOHLMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service