Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W386901

Sigma-Aldrich

3-Methyl-2-oxobutanoic acid sodium salt

95%

Synonym(s):

Sodium 3-methyl-2-oxobutyrate, α-Ketoisovaleric acid sodium salt, 2-Keto-3-methylbutyric acid sodium salt, 3-Methyl-2-oxobutanoic acid sodium salt, 3-Methyl-2-oxobutyric acid sodium salt, Ketovaline sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCOONa
CAS Number:
Molecular Weight:
138.10
Beilstein/REAXYS Number:
4334552
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

assay

95%

mp

220-230 °C (dec.) (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fruity

SMILES string

[Na+].CC(C)C(=O)C([O-])=O

InChI

1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1

InChI key

WIQBZDCJCRFGKA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix S Krause et al.
Applied and environmental microbiology, 76(24), 8053-8061 (2010-10-12)
2-Ketoisovalerate is used as a therapeutic agent, and a 2-ketoisovalerate-producing organism may serve as a platform for products deriving from this 2-keto acid. We engineered the wild type of Corynebacterium glutamicum for the growth-decoupled production of 2-ketoisovalerate from glucose by
Marta de la Plaza et al.
Journal of molecular microbiology and biotechnology, 17(2), 96-100 (2008-11-27)
The alpha-ketoisovalerate decarboxylase (Kivd) is a unique lactococcal key enzyme in the decarboxylation of branched-chain alpha-keto acids derived from branched-chain amino acids transamination into aldehydes. These products are important aroma compounds produced during cheese ripening. In this study, the Kivd
Taro Murakami et al.
Journal of nutritional science and vitaminology, 51(1), 48-50 (2005-05-27)
Branched-chain alpha-keto acid dehydrogenase kinase (BDK) phosphorylates and inactivates the branched-chain alpha-keto acid dehydrogenase complex (BCKDC), which is the rate-limiting enzyme in the branched-chain amino acid catabolism. BDK has been believed to be bound to the BCKDC. However, recent our
Isabel Ayala et al.
Journal of biomolecular NMR, 43(2), 111-119 (2008-12-31)
A strategy for the introduction of ((1)H,(13)C-methyl)-alanine into perdeuterated proteins is described. Specific protonation of alanine methyl groups to a level of 95% can be achieved by overexpressing proteins in M9/D(2)O based bacterial growth medium supplemented with 800 mg/l of
Benjamin F Miller et al.
The Journal of physiology, 567(Pt 3), 1021-1033 (2005-07-09)
We hypothesized that an acute bout of strenuous, non-damaging exercise would increase rates of protein synthesis of collagen in tendon and skeletal muscle but these would be less than those of muscle myofibrillar and sarcoplasmic proteins. Two groups (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service