Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W369705

Sigma-Aldrich

2-Hydroxy-4-methylbenzaldehyde

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(CH3)CHO
CAS Number:
Molecular Weight:
136.15
FEMA Number:
3697
EC Number:
Council of Europe no.:
2130
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.091
NACRES:
NA.21
organoleptic:
almond
grade:
Kosher
biological source:
synthetic
agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

agency

meets purity specifications of JECFA

assay

≥98%

mp

58-61 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

almond

SMILES string

[H]C(=O)c1ccc(C)cc1O

InChI

1S/C8H8O2/c1-6-2-3-7(5-9)8(10)4-6/h2-5,10H,1H3

InChI key

JODRRPJMQDFCBJ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service