Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W314005

Sigma-Aldrich

2,6-Dimethyl-4-heptanol

≥82%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CH3)CH2CH(OH)CH2CH(CH3)CH3
CAS Number:
Molecular Weight:
144.25
FEMA Number:
3140
EC Number:
Council of Europe no.:
11719
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.081
Pricing and availability is not currently available.

biological source

synthetic

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

assay

≥82%

impurities

<18% 4,6-Dimethyl-2-heptanol

refractive index

n20/D 1.423 (lit.)

bp

178 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

fragrance allergen

no known allergens

SMILES string

CC(C)CC(O)CC(C)C

InChI

1S/C9H20O/c1-7(2)5-9(10)6-8(3)4/h7-10H,5-6H2,1-4H3

InChI key

HXQPUEQDBSPXTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

150.8 °F - closed cup

flash_point_c

66 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service