Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W287601

Sigma-Aldrich

Phenylacetaldehyde dimethyl acetal

≥98%, FCC

Synonym(s):

(2,2-Dimethoxyethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(OCH3)2
CAS Number:
Molecular Weight:
166.22
FEMA Number:
2876
Beilstein/REAXYS Number:
879360
EC Number:
Council of Europe no.:
40
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.006
NACRES:
NA.21

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FCC

assay

≥98%

refractive index

n20/D 1.493 (lit.)

bp

219-221 °C/754 mmHg (lit.)

density

1.004 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; floral

SMILES string

COC(Cc1ccccc1)OC

InChI

1S/C10H14O2/c1-11-10(12-2)8-9-6-4-3-5-7-9/h3-7,10H,8H2,1-2H3

InChI key

WNJSKZBEWNVKGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

192.2 °F - closed cup

flash_point_c

89 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service