Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W225118

Sigma-Aldrich

(−)-Carvyl propionate

≥97%

Synonym(s):

(−)-Carvyl propionate, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O2
CAS Number:
Molecular Weight:
208.30
FEMA Number:
2251
EC Number:
Council of Europe no.:
424
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.143

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

assay

≥97%

optical activity

[α]20/D −107°, neat

refractive index

n20/D 1.474 (lit.)
n20/D 1.474

bp

78-80 °C/0.2 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

minty; sweet

SMILES string

CCC(=O)OC1CC(CC=C1C)C(C)=C

InChI

1S/C13H20O2/c1-5-13(14)15-12-8-11(9(2)3)7-6-10(12)4/h6,11-12H,2,5,7-8H2,1,3-4H3

InChI key

DFVXNZOMAOGTBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service