Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W221055

Sigma-Aldrich

Isobutyl phenylacetate

natural, ≥98%, FG

Synonym(s):

Phenyl acetic acid isobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
192.25
FEMA Number:
2210
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.788
organoleptic:
cocoa; honey; floral; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

botanical

grade

FG
Fragrance grade
Halal
Kosher
natural

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

assay

≥98%

refractive index

n20/D 1.487 (lit.)

bp

253 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

cocoa; honey; floral; sweet

SMILES string

CC(C)COC(=O)Cc1ccccc1

InChI

1S/C12H16O2/c1-10(2)9-14-12(13)8-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

RJASFPFZACBKBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service