Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W209890

Sigma-Aldrich

Anisyl acetate

natural, 97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2C6H4-4-(OCH3)
CAS Number:
Molecular Weight:
180.20
FEMA Number:
2098
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.019
NACRES:
NA.21
organoleptic:
anise; cherry; sweet; vanilla
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

97%

refractive index

n20/D 1.513 (lit.)

bp

137-139 °C/12 mmHg (lit.)

density

1.107 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

anise; cherry; sweet; vanilla

SMILES string

COc1ccc(COC(C)=O)cc1

InChI

1S/C10H12O3/c1-8(11)13-7-9-3-5-10(12-2)6-4-9/h3-6H,7H2,1-2H3

InChI key

HFNGYHHRRMSKEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service