Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P46156

Sigma-Aldrich

4-Piperidineethanol

97%

Synonym(s):

4-(2-Hydroxyethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

refractive index

n20/D 1.4902 (lit.)

bp

131-136 °C/17 mmHg (lit.)

mp

46-47 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCC1CCNCC1

InChI

1S/C7H15NO/c9-6-3-7-1-4-8-5-2-7/h7-9H,1-6H2

InChI key

LDSQQXKSEFZAPE-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Ursolic acid derivatives
  • Amodiaquine analogues for treatment of cerebral malaria
  • Spiroimidazolidinone NPC1L1 inhibitors
  • Piperazinyl glutamate pyridines as orally bioavailable P2Y12 antagonists for inhibition of platelet aggregation
  • Neurokinin-2 receptor antagonists
  • Hepatitis C virus NS5B polymerase inhibitors

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service