Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I22102

Sigma-Aldrich

N-Isopropylethylenediamine

98%

Synonym(s):

2-(Isopropylamino)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH2CH2NH2
CAS Number:
Molecular Weight:
102.18
Beilstein/REAXYS Number:
1732183
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)

density

0.819 g/mL at 25 °C (lit.)

SMILES string

CC(C)NCCN

InChI

1S/C5H14N2/c1-5(2)7-4-3-6/h5,7H,3-4,6H2,1-2H3

InChI key

KDRUIMNNZBMLJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

42.8 °F - closed cup

flash_point_c

6 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service