Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H31344

Sigma-Aldrich

4-Hydroxy-9-fluorenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

98%

SMILES string

Oc1cccc2C(=O)c3ccccc3-c12

InChI

1S/C13H8O2/c14-11-7-3-6-10-12(11)8-4-1-2-5-9(8)13(10)15/h1-7,14H

InChI key

OZLKIIAMQWXVKQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service