Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F5381

Sigma-Aldrich

Iron(II) fumarate

≥95%

Synonym(s):

Ferrous fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2FeO4
CAS Number:
Molecular Weight:
169.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

SMILES string

O=C1O[Fe]OC(=O)\C=C\1

InChI

1S/C4H4O4.Fe/c5-3(6)1-2-4(7)8;/h1-2H,(H,5,6)(H,7,8);/q;+2/p-2/b2-1+;

InChI key

PMVSDNDAUGGCCE-TYYBGVCCSA-L

Looking for similar products? Visit Product Comparison Guide

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pilar Garrido-Martín et al.
Interactive cardiovascular and thoracic surgery, 15(6), 1013-1018 (2012-09-04)
Anaemia is a frequent complication after cardiopulmonary bypass surgery. Iron therapy has been variably employed by medical centres over the years. In our study we test the clinical effectiveness of intravenous and oral iron supplementation in correcting anaemia, and its
Divya Yadava et al.
Journal of microencapsulation, 29(8), 729-738 (2012-08-10)
An extrusion-based encapsulation process has been developed for making salt grain-sized iron premix for salt fortification. The first step of extrusion agglomeration process has been studied and reported previously. The focus of this study is on the optimisation of the
Yao Olive Li et al.
Journal of microencapsulation, 28(7), 639-649 (2011-08-10)
A microencapsulation-based technology platform has been developed for salt double fortification with iron and iodine, aiming to address two globally prevalent micronutrient deficiencies simultaneously. Specifically, ferrous fumarate was microencapsulated into a form of salt grain-sized premix, and then added into
eComment. The problem of anaemia correction in cardiac surgery patients.
Leo A Bockeria et al.
Interactive cardiovascular and thoracic surgery, 15(6), 1018-1018 (2012-11-21)
M Harrington et al.
European journal of clinical nutrition, 65(1), 20-25 (2010-09-16)
Ferrous fumarate is recommended for the fortification of complementary foods based on similar iron absorption to ferrous sulfate in adults. Two recent studies in young children have reported that it is only 30% as well absorbed as ferrous sulfate. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service