Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E30751

Sigma-Aldrich

Ethyl 2-hydroxyethyl sulfide

97%

Synonym(s):

2-(Ethylthio)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SCH2CH2OH
CAS Number:
Molecular Weight:
106.19
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

refractive index

n20/D 1.4869 (lit.)

bp

180-184 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCSCCO

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

172.4 °F

flash_point_c

78 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service