Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS003174

Sigma-Aldrich

4-Hydroxypyrrolidine-2-carboxylic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

OC1CNC(C1)C(O)=O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)

Inchi Key

PMMYEEVYMWASQN-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryce Sunsdahl et al.
Angewandte Chemie (International ed. in English), 53(52), 14352-14356 (2014-10-11)
Highly diastereoselective metalloamination/cyclization reactions of zinc(II) hydrazides obtained through reaction of diethylzinc with N,N-dimethylhydrazinoalkenes are described. The resulting organozinc intermediates undergo facile allylation and acylation, in situ, to provide the corresponding functionalized piperidines and pyrrolidines.
Flavia Emi Akamatsu et al.
Histology and histopathology, 31(5), 515-522 (2015-09-05)
We investigated the action of radiofrequency (RF) on the healing process after inducing experimental lesions of the Achilles tendon in rats. Wistar rats were surgically subjected to bilateral partial transverse sectioning of the Achilles tendon. The right tendon was treated
Ozlem Aydin et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(6), 1238-1248 (2014-10-29)
Osteoarthritis (OA) is a degenerative joint disease, which has no complete treatment with medication yet. Intraarticular hyaluronan (HA) injection can decrease pain and modify the natural course of OA. This study was designed to provide long term delivery of an
Min Kim et al.
Experimental & molecular medicine, 52(10), 1766-1776 (2020-10-29)
Non-alcoholic fatty liver disease (NAFLD) is the most common liver disease and can lead to multiple complications, including non-alcoholic steatohepatitis (NASH), cirrhosis, and hepatocellular carcinoma. The fibrotic liver is characterized by the pathological accumulation of extracellular matrix (ECM) proteins. Type
Carina I Mack et al.
Molecular nutrition & food research, 64(9), e1901190-e1901190 (2020-03-15)
Diabetes is thought to primarily represent a disturbance of carbohydrate metabolism; however, population studies employing metabolomics have mainly identified plasma amino acids and lipids, or their products, as biomarkers. In this pilot study, the aim is to analyze a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service