Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS000527

Sigma-Aldrich

Propyl salicylate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O3
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CCCOC(=O)c1ccccc1O

InChI

1S/C10H12O3/c1-2-7-13-10(12)8-5-3-4-6-9(8)11/h3-6,11H,2,7H2,1H3

InChI key

LZFIOSVZIQOVFW-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soliman Mohammadi-Samani et al.
Burns : journal of the International Society for Burn Injuries, 42(2), 428-433 (2016-01-21)
This study was designed to investigate the effect of inorganic phosphate supplementation on invasive behavior of Pseudomonas aeruginosa in burn wound infections. An emulsion-based lotion containing sodium dihydrogen phosphate was formulated and then 50 female Sprague-Dawley rats with burn wounds
Srinivas Agurla et al.
Plant physiology and biochemistry : PPB, 157, 276-283 (2020-11-06)
Modulation by salicylic acid (SA) and its six esters of stomatal closure was evaluated in Arabidopsis thaliana. The seven compounds tested are salicylic acid (SA), acetylsalicylate (ASA), methyl salicylate (MeSA), propyl salicylate (PrSA), amyl salicylate, benzyl salicylate, and salicin. Among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service