Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A56108

Sigma-Aldrich

Aminoguanidine nitrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHC(=NH)NH2 · HNO3
CAS Number:
Molecular Weight:
137.10
Beilstein/REAXYS Number:
3916056
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

crystals

mp

145-147 °C (lit.)

SMILES string

O[N+]([O-])=O.NNC(N)=N

InChI

1S/CH6N4.HNO3/c2-1(3)5-4;2-1(3)4/h4H2,(H4,2,3,5);(H,2,3,4)

InChI key

PMGFHEJUUBDCLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service