Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W515701

Sigma-Aldrich

Magnesium phosphate hydrate

≥98% (ignited basis)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg3(PO4)2·xH2O
CAS Number:
Molecular Weight:
262.86 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

assay

≥98% (ignited basis)

density

2.195 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

faint

SMILES string

O.[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/3Mg.2H3O4P.H2O/c;;;2*1-5(2,3)4;/h;;;2*(H3,1,2,3,4);1H2/q3*+2;;;/p-6

InChI key

WESIJKDWGUWFEP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium phosphate hydrate finds application as a nutritional supplement in foods.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannai S
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 819-819 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service