Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W432900

Sigma-Aldrich

2,4,6-Trimethylphenol

99%, FG

Synonym(s):

Mesitol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
4329
Beilstein/REAXYS Number:
1859675
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.095
organoleptic:
coffee; phenolic
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

assay

99%

bp

220 °C (lit.)

mp

70-72 °C (lit.)
71-74 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

coffee; phenolic

SMILES string

Cc1cc(C)c(O)c(C)c1

InChI

1S/C9H12O/c1-6-4-7(2)9(10)8(3)5-6/h4-5,10H,1-3H3

InChI key

BPRYUXCVCCNUFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

target_organs

Respiratory system, Stomach

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieqiong Wang et al.
Water research, 173, 115524-115524 (2020-02-02)
Nebivolol (NEB) is widely used for the treatment of hypertension and chronic heart failure and has become an ubiquitous emerging organic pollutant. It has been shown to undergo direct photolysis, but the role of DOM in its degradation kinetics and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service