Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W361704

Sigma-Aldrich

Benzyl disulfide

98%, FG

Synonym(s):

Dibenzyl disulfide, Benzyl disulfide, Bis(phenylmethyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SSCH2C6H5
CAS Number:
Molecular Weight:
246.39
FEMA Number:
3617
Beilstein/REAXYS Number:
1110443
EC Number:
Council of Europe no.:
4077
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.081
NACRES:
NA.21
organoleptic:
green; earthy; smoky
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

assay

98%

mp

69-72 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; earthy; smoky

SMILES string

C(SSCc1ccccc1)c2ccccc2

InChI

1S/C14H14S2/c1-3-7-13(8-4-1)11-15-16-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

GVPWHKZIJBODOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Munday et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(11), 1609-1615 (2003-09-10)
Thiols and disulphides make an important contribution to the organoleptic characteristics of foodstuffs, and many such compounds are approved for use as food flavours. Some thiols and disulphides are haemolytic agents in vivo. The haemolysis is caused by oxidative damage
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service