Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W265713

Sigma-Aldrich

Mandarin oil

natural, Italian, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2657
Council of Europe no.:
142
MDL number:
UNSPSC Code:
12164502

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.4745

bp

178 °C (lit.)

density

0.846 g/mL at 25 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

orange; floral; citrus

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Mandarin peel.

Biochem/physiol Actions

Taste at 5 ppm

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

133.0 °F - closed cup

flash_point_c

56.11 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 28, 78-78 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service