Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W241601

Sigma-Aldrich

Ethyl 2-acetyl-3-phenylpropionate

≥97%

Synonym(s):

Ethyl 2-benzylacetoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(CH2C6H5)CO2C2H5
CAS Number:
Molecular Weight:
220.26
FEMA Number:
2416
EC Number:
Council of Europe no.:
2241
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.501

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

assay

≥97%

refractive index

n20/D 1.5 (lit.)
n20/D 1.5

bp

276 °C (lit.)
276 °C

density

1.036 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

balsam; jasmine; fruity

SMILES string

CCOC(=O)C(Cc1ccccc1)C(C)=O

InChI

1S/C13H16O3/c1-3-16-13(15)12(10(2)14)9-11-7-5-4-6-8-11/h4-8,12H,3,9H2,1-2H3

InChI key

XDWQYMXQMNUWID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service