Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W222909

Sigma-Aldrich

(+)-Camphene

≥90%, FCC

Synonym(s):

(1R)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2229
EC Number:
Council of Europe no.:
2227
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.009

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

assay

≥90%

optical activity

[α]20/D +16°, c = 4 in ethanol

bp

159-160 °C (lit.)

mp

40-50 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

camphoraceous

SMILES string

CC1(C)[C@@H]2CC[C@@H](C2)C1=C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m0/s1

InChI key

CRPUJAZIXJMDBK-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 100 ppm

Other Notes

Natural occurrence: Carrot, dill, fennel, tarragon, marjoram, thyme, nutmeg, parsley and pepper.

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 53-53 (2005)
Mosciano, G.
Perfumer & Flavorist, 27, 70-70 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service