Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

97230

Sigma-Aldrich

Z-Thr-OH

≥98.5% (T)

Synonym(s):

N-Carbobenzyloxy-L-threonine, N-Cbz-L-threonine, Z-L-Threonine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
253.25
Beilstein/REAXYS Number:
2335409
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (T)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

101-103 °C (lit.)

application(s)

peptide synthesis

SMILES string

C[C@@H](O)[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C12H15NO5/c1-8(14)10(11(15)16)13-12(17)18-7-9-5-3-2-4-6-9/h2-6,8,10,14H,7H2,1H3,(H,13,17)(H,15,16)/t8-,10+/m1/s1

InChI key

IPJUIRDNBFZGQN-SCZZXKLOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service