Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82685

Sigma-Aldrich

3(2H)-Pyridazinone

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O
CAS Number:
Molecular Weight:
96.09
Beilstein/REAXYS Number:
106974
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (GC)

form

solid

impurities

≤4% water

mp

98-104 °C

SMILES string

O=C1NN=CC=C1

InChI

1S/C4H4N2O/c7-4-2-1-3-5-6-4/h1-3H,(H,6,7)

InChI key

AAILEWXSEQLMNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuhiro Terada et al.
Neurology, 94(15), e1592-e1604 (2020-03-07)
In vivo glycolysis-related glucose metabolism and electron transport chain-related mitochondrial activity may be different regionally in the brains of patients with Alzheimer disease (AD). To test this hypothesis regarding AD pathophysiology, we measured the availability of mitochondrial complex-I (MC-I) with
Jan Chabera et al.
Ecotoxicology and environmental safety, 208, 111645-111645 (2021-01-06)
The effects of chloridazon (Ch) and its metabolite chloridazon-desphenyl (Ch-D) at the environmentally relevant concentrations of 0.45 µg/L and 2.7 µg/L on signal crayfish Pacifastacus leniusculus were assessed in a 30-day exposure followed by a 15-day depuration period. Locomotion, biochemical haemolymph profile
Hanna Rostkowska et al.
The journal of physical chemistry. A, 121(37), 6932-6941 (2017-08-30)
The UV-induced thione → thiol phototautomeric reaction has been studied for monomeric 3-thio-1,2,4-triazole (3-ST) isolated in low-temperature Ar and n-H
Hawazen BinMahfouz et al.
Molecular pharmacology, 87(1), 64-76 (2014-10-18)
Glucocorticoids, also known as corticosteroids, induce effector gene transcription as a part of their anti-inflammatory mechanisms of action. Such genomic effects can be significantly enhanced by long-acting β2-adrenoceptor agonists (LABAs) and may contribute to the clinical superiority of inhaled corticosteroid
Natália Corniani et al.
PloS one, 9(7), e103704-e103704 (2014-08-01)
The 2-C-methyl-D-erythritol 4-phosphate (MEP) pathway leads to the synthesis of isopentenyl diphosphate in plastids. It is a major branch point providing precursors for the synthesis of carotenoids, tocopherols, plastoquinone and the phytyl chain of chlorophylls, as well as the hormones

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service