Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

776432

Sigma-Aldrich

Polystyrene-block-poly(tert-butyl acrylate), DDMAT terminated, acid terminated

average Mn 12,000, block ratio 50:50, PDI ≤1.2

Synonym(s):

PS(50)-b-PtBA(50), RAFT poly(styrene-block-tbutyl acrylate) carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

mol wt

average Mn 12,000

transition temp

Tm 245 °C

PDI

≤1.2

Application

This is a well-defined diblock copolymer containing a polystyrene block which is hydrophobic; and a poly(tert-butyl acrylate) which is hydrophobic. This polystyrene-block-poly(t-Butylacrylate) polymer is used in many applications; primarily in which the tert-butyl group is substituted with a desired functionality. The hydrophilicity of this t-butyl acrylate block can be tuned by varying the extent of hydrolysis of the t-butyl groups. The end group functionality contains a carboxylic acid (COOH) as well as the Dodecyltrithiocarbonate RAFT agent moiety. This RAFT CTA will allow for additional polymerization to generate a diblock copolymer and is especially suited for the polymerization of styrene; acrylate and acrylamide monomers. It could also be hydrolyzed to form a different funcitonality.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service