Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

772887

Sigma-Aldrich

Lanthanum borate

powder, -200 mesh, 99.9% trace metals basis

Synonym(s):

Boron lanthanum oxide, Lanthanum orthoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaBO3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

particle size

-200 mesh

mp

>300 °C

SMILES string

[La+3].[O-]B([O-])[O-]

InChI

1S/BO3.La/c2-1(3)4;/q-3;+3

InChI key

GJJSDZSDOYNJSW-UHFFFAOYSA-N

application

LaBO3 is a promising phosphor host material, showing different emission properties upon doping with rare earth and other elements. LaBO3 doped with alkaline and other metals are also studied as solid state proton conductors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahashi, H.
Solid State Ionics, 192, 275-275 (2011)
Jubera, V.
Journal of Luminescence, 101, 1-1 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service