74295
3-Nonanol
≥95.0% (GC)
Synonym(s):
Ethyl hexyl carbinol
About This Item
assay
≥95.0% (GC)
refractive index
n20/D 1.431
bp
192-194 °C (lit.)
density
0.824 g/mL at 20 °C (lit.)
SMILES string
CCCCCCC(O)CC
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
204.8 °F - closed cup
flash_point_c
96 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service