735930
Zinc fluoride
anhydrous, 99.999% trace metals basis
About This Item
grade
anhydrous
vapor pressure
1 mmHg ( 970 °C)
assay
99.999% trace metals basis
mp
>400 °C
872 °C (lit.)
density
4.95 g/mL at 25 °C (lit.)
SMILES string
F[Zn]F
InChI
1S/2FH.Zn/h2*1H;/q;;+2/p-2
InChI key
BHHYHSUAOQUXJK-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service