Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706515

Sigma-Aldrich

Copper hydride (CuH) solution

in toluene

Synonym(s):

"Hot" Stryker′s Reagent, (BDP)Cu(H), Copper monohydride, Cuprous hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuH
CAS Number:
Molecular Weight:
64.55
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: copper
reagent type: catalyst

concentration

in toluene

storage temp.

−20°C

SMILES string

[CuH]

InChI

1S/Cu.H

InChI key

JJFLDSOAQUJVBF-UHFFFAOYSA-N

Application

Reagent for stereoselective synthesis of exocyclic tetra substituted enol ethers and olefins

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

42.8 °F

flash_point_c

6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niklas O Thiel et al.
Organic & biomolecular chemistry, 14(45), 10660-10666 (2016-10-27)
An air-stable and preactivated copper(i) hydroxide/N-heteroyclic carbene (NHC) complex for alkyne semihydrogenations is reported. Next to an enhanced practicability of the process, the resulting alkenes are obtained with high Z-selectivities and no overreduction to the corresponding alkanes.
Benjamin A Baker et al.
Organic letters, 10(2), 289-292 (2007-12-21)
A ligand-modified, economical version of Stryker's reagent (SR) has been developed based on a bidentate, achiral bis-phosphine. Generated in situ, "(BDP)CuH" smoothly effects conjugate reductions of a variety of unsaturated substrates, including those that are normally unreactive toward SR. Substrate-to-ligand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service