Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687154

Sigma-Aldrich

1-Boc-2-aminomethylpiperidine

95%

Synonym(s):

1,1-Dimethylethyl 2-(aminomethyl)-1-piperidinecarboxylate, 2-Aminomethylpiperidine-1-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.4745

density

1.0124 g/mL at 25 °C

functional group

amine

SMILES string

CC(C)(C)OC(=O)N1CCCCC1CN

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-7-5-4-6-9(13)8-12/h9H,4-8,12H2,1-3H3

InChI key

PTVRCUVHYMGECC-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • TRPV1 antagonists
  • CHK1 inhibitors
  • p38a MAP kinase inhibitors
  • Carbamoylphosphonates
  • Macrolactams via carbonylation/intramolecular amidation
  • Intramolecular transamidation

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service