Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

686816

Sigma-Aldrich

5-Bromopyridine-3-boronic acid

≥95%

Synonym(s):

(3-Bromopyridin-5-yl)boronic acid, (5-Bromo-3-pyridyl)boronic acid, (5-bromo-3-pyridinyl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BBrNO2
CAS Number:
Molecular Weight:
201.81
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

powder

mp

260 (dec.)

storage temp.

2-8°C

SMILES string

OB(O)c1cncc(Br)c1

InChI

1S/C5H5BBrNO2/c7-5-1-4(6(9)10)2-8-3-5/h1-3,9-10H

InChI key

ICCGFOKNFZWCTJ-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant for preparation of:
  • Thienylpyridyl garlands via cross-coupling with heteroaryl halides
  • Arenes via nickel-catalyzed cross-coupling with potassium aryl- and heteroaryl trifluoroborates
  • Meso-substituted ABCD-type porphyrins by functionalization reactions
  • Analogs of (acetylamino)(dimethylpyrazol)(pyridyl)pyrimidines as A2A adenosine receptor antagonists for the treatment of Parkinson′s disease
  • 5-hydroxy-1,3,2-dioxaborolan-4-ones boronate-amine complexes via heterocyclization with α-oxocarboxylic acids followed by subsequent complexation
  • Radioligands with optimized brain kinetics for PET imaging of nAChR

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service