Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683299

Sigma-Aldrich

5′-Fluoro-2′-methoxyacetophenone

97%

Synonym(s):

1-(5-Fluoro-2-methoxyphenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9FO2
CAS Number:
Molecular Weight:
168.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.5186

density

1.1880 g/mL at 25 °C

functional group

fluoro
ketone

SMILES string

COc1ccc(F)cc1C(C)=O

InChI

1S/C9H9FO2/c1-6(11)8-5-7(10)3-4-9(8)12-2/h3-5H,1-2H3

InChI key

CNVGMLMIQIPAFY-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service