Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67778

Sigma-Aldrich

Methanethiol

Synonym(s):

Mercaptan C1, Methyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SH
CAS Number:
Molecular Weight:
48.11
Beilstein/REAXYS Number:
1696840
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Product 67778 is not currently sold in your country. Contact Technical Service

vapor density

1.66 (vs air)

Quality Level

vapor pressure

1536 mmHg ( 20 °C)

expl. lim.

21.8 %

bp

6 °C (lit.)

mp

−123 °C (lit.)

functional group

thiol

storage temp.

2-8°C

SMILES string

CS

InChI

1S/CH4S/c1-2/h2H,1H3

InChI key

LSDPWZHWYPCBBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 25 mL (net 22 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

-0.4 °F - closed cup

flash_point_c

-18 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yaegaki et al.
Journal of periodontal research, 27(4 Pt 1), 233-238 (1992-07-01)
Volatile sulfur compounds (VSC) in mouth air were estimated by gas chromatography. The amount of VSC and the methyl mercaptan/hydrogen sulfide ratio were significantly increased in patients with periodontal disease. These two parameters also increased in proportion to the bleeding
N J Hayward et al.
Journal of clinical microbiology, 6(3), 195-201 (1977-09-01)
Proteus mirabilis was distinguishable from Escherichia coli and from several other species that may be associated with urinary tract infections when grown in a nutrient medium supplemented with 0.1 M L-methionine by the formation of large amounts of dimethyl disulfide
Ari Zeida et al.
Chemical research in toxicology, 25(3), 741-746 (2012-02-07)
The oxidation of cellular thiol-containing compounds, such as glutathione and protein Cys residues, is considered to play an important role in many biological processes. Among possible oxidants, hydrogen peroxide (H(2)O(2)) is known to be produced in many cell types as
Govindan Muthuraman et al.
Journal of hazardous materials, 193, 257-263 (2011-08-16)
In this study, an electrocatalytic wet scrubbing process was developed for the simultaneous removal of synthetic odorous gases namely, methyl mercaptan (CH(3)SH) and hydrogen sulfide (H(2)S). The initial process consists of the absorption of CH(3)SH and H(2)S gases by an
Kazue Yamagishi et al.
Gut, 61(4), 554-561 (2011-08-13)
The mechanisms of cancer cell growth and metastasis are still not entirely understood, especially from the viewpoint of chemical reactions in tumours. Glycolytic metabolism is markedly accelerated in cancer cells, causing the accumulation of glucose (a reducing sugar) and methionine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service