Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

674036

Sigma-Aldrich

3-Bromo-2-hydroxybenzaldehyde

97%

Synonym(s):

3-Bromosalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrO2
CAS Number:
Molecular Weight:
201.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

53-57 °C

SMILES string

[H]C(=O)c1cccc(Br)c1O

InChI

1S/C7H5BrO2/c8-6-3-1-2-5(4-9)7(6)10/h1-4,10H

InChI key

STBGLXMINLWCNL-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Guo-Ping Cheng et al.
Acta chimica Slovenica, 64(1), 261-265 (2017-04-06)
A rarely seen phenolato and azido-bridged polymeric cadmium(II) complex derived from the Schiff base ligand 2-bromo-6-[(2-isopropylaminoethylimino)methyl]phenol (HL) has been prepared and characterized by elemental analysis, IR spectroscopy, and single crystal X-ray diffraction. The Schiff base ligand coordinates to the Cd
Shao-Song Qian et al.
Acta chimica Slovenica, 62(4), 828-833 (2015-12-19)
Reaction of [VO(acac)(2)] (where acac = acetylacetonate), benzohydroxamic acid (Hbha), and two similar aroylhydrazone ligands in methanol produced two benzohydroxamate-coordinated mononuclear vanadium(V) oxo complexes with general formula [VOL(bha)], where L = L(1) = N'-(5-bromo-2-hydroxybenzylidene)-2-fluorobenzohydrazide (H(2)L(1)), and L = L(2) =
Florian Juhlke et al.
Frontiers in chemistry, 5, 120-120 (2018-01-13)
Chlorinated guaiacol derivatives are found in waste water of pulp mills using chlorine in the bleaching process of wood pulp. They can also be detected in fish tissue, possibly causing off-odors. To date, there is no systematic investigation on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service