668028
Methyl 5-methyl-2-furoate
97%
Synonym(s):
5-Methylfuran-2-carboxylic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
assay
97%
refractive index
n20/D 1.493
bp
255 °C
density
1.114 g/mL at 25 °C
SMILES string
COC(=O)c1ccc(C)o1
InChI
1S/C7H8O3/c1-5-3-4-6(10-5)7(8)9-2/h3-4H,1-2H3
InChI key
XBYZJUMTKHUJIY-UHFFFAOYSA-N
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
189.0 °F - closed cup
flash_point_c
87.2 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service