Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

666912

Sigma-Aldrich

4-Methoxy-2-(trifluoromethyl)phenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)C6H3(CF3)B(OH)2
CAS Number:
Molecular Weight:
219.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

solid

mp

168-172 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

COc1ccc(B(O)O)c(c1)C(F)(F)F

InChI

1S/C8H8BF3O3/c1-15-5-2-3-7(9(13)14)6(4-5)8(10,11)12/h2-4,13-14H,1H3

InChI key

ZBCRZEJNAADYKG-UHFFFAOYSA-N

application

Reactant involved in:
  • Synthesis of pyrazine derivatives as corticotropin releasing factor-1 receptor antagonists
  • C-H functionalization of quinones
  • Phenyl-purine-carbonitrile derivative synthesis as cathepsin S inhibitors
  • Synthesis of chiral bicyclooctadiene-based ligands
  • Synthesis of corticotropin-releasing factor-1 antagonists

Other Notes

May contain varying amounts of anhydride

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service