Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660191

Sigma-Aldrich

S,S′-Dimethyl dithiocarbonate

≥96%

Synonym(s):

Carbonodithioic acid S,S-dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6S2O
CAS Number:
Molecular Weight:
122.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥96%

refractive index

n20/D 1.5485

density

1.1705 g/mL at 25 °C

functional group

thioether

SMILES string

CSC(=O)SC

InChI

1S/C3H6OS2/c1-5-3(4)6-2/h1-2H3

InChI key

IUXMJLLWUTWQFX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service