All Photos(1)
About This Item
Linear Formula:
(CH2CH(C6H5))x(CH2CH(C7H6Cl))y
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
General description
1:1 mole ratio of styrene:chloromethylstyrene
Features and Benefits
Polymer will self-assemble into nanocapsules for drug encapsulation and drug delivery applications.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shenhar, R. et al.
Advanced Materials, 17, 657-657 (2005)
Deans, R. et al.
Macromolecules, 32, 4956-4956 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service