Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

653683

Sigma-Aldrich

5-Nitro-2-oxindole

97%

Synonym(s):

5-Nitro-1,3-dihydroindole-2-one, NSC 25199, NSC 99066

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O3
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

233-236 °C

SMILES string

[O-][N+](=O)c1ccc2NC(=O)Cc2c1

InChI

1S/C8H6N2O3/c11-8-4-5-3-6(10(12)13)1-2-7(5)9-8/h1-3H,4H2,(H,9,11)

InChI key

JQCGHRDKVZPCRO-UHFFFAOYSA-N

Application

  • Reactant for synthesis of 3-[4-(amino/methylsulfonyl)phenyl]methylene-indolin-2-one derivatives as novel COX-1/2 and 5-LOX inhibitors
  • Reactant for preparation of Aurora kinase inhibitors
  • Reactant for synthesis of 3-substituted 2-indolinone RET inhibitors
  • Reactant for synthesis of 4-azachromeno[2,3-b]indol-11(6H)-ones and derivatives as analogs of ellipticine
  • Reactant for preparation of [18F]fluorobenzylidene-indolinone as possible tyrosine kinase inhibitor for PET tumor imaging
  • Reactant for preparation of 3-substituted indolin-2-ones as neuroprotective and toxic agents

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Letribot et al.
Molecules (Basel, Switzerland), 23(6) (2018-06-13)
Alkylidene oxindoles are important functional moieties and building blocks in pharmaceutical and synthetic chemistry. Our interest in biologically active compounds focused our studies on the synthesis of novel oxindoles, bearing on the exocyclic double bond at the C8, CN, and
Ayman M Saleh et al.
Molecules (Basel, Switzerland), 19(9), 13076-13092 (2014-08-27)
A selected set of substituted pyridone-annelated isoindigos 3a-f has been synthesized via interaction of 5- and 6-substituted oxindoles 2a-f with 6-ethyl-1,2,9-trioxopyrrolo[3,2-f]quinoline-8-carboxylic acid (1) in acetic acid at reflux. Among these isoindigos, the 5'-chloro and 5'-bromo derivatives 3b and 3d show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service