Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636266

Sigma-Aldrich

5-Amino-2,2-dimethylpentanol

Synonym(s):

2,2-Dimethyl-5-amino-1-pentanol, 5-Amino-2,2-dimethyl-1-pentanol, 5-Amino-2,2-dimethyl-1-pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO
CAS Number:
Molecular Weight:
131.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

CC(C)(CO)CCCN

InChI

1S/C7H17NO/c1-7(2,6-9)4-3-5-8/h9H,3-6,8H2,1-2H3

InChI key

ZOOPONUAQQEUQX-UHFFFAOYSA-N

Application

Reactant for:
  • Reductive amination with borohydride and borane reducing agents
  • Copper-catalyzed N- and O-arylation with aryl iodides
  • Preparation of N-(alkyl)arylamine derivatives via selective room-temperature carbon-nitrogen coupling reaction using copper iodide as catalyst and diketone as ligand

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service