Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

607010

Sigma-Aldrich

Dichloromethane-12C,d2

99.9 atom % D, 99.95 atom % 12C

Synonym(s):

Methylene chloride-12C,d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CD2Cl2
Molecular Weight:
86.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

description

13C depleted

isotopic purity

99.95 atom % 12C
99.9 atom % D

assay

99% (CP)

bp

40 °C (lit.)

mp

-97 °C (lit.)

density

1.356 g/mL at 25 °C

application(s)

agriculture

mass shift

M+2

SMILES string

[2H][12C]([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1+0D2

InChI key

YMWUJEATGCHHMB-WUVOJTTISA-N

General description

Dichloromethane-12C,d2 (12CD2Cl2) is a methylene chloride isotopomer. High resolution infrared spectra of 12CH2Cl2, 13CH2Cl2, and 12CD2Cl2 in the gas phase have been recorded.

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylene chloride: The mid-infrared spectrum of an almost vibrationally unperturbed molecule.
Duncan JL, et al.
Journal of Molecular Spectroscopy, 118(1), 145-162 (1986)
Ting Li et al.
Environmental science and pollution research international, 22(15), 11792-11800 (2015-04-11)
We surveyed the occurrence of nine N-nitrosamine species in ten bottled drinking waters from supermarket and other water samples including raw waters, finished waters, and distribution system waters from nine municipal drinking water treatment plants in eight cities of Jiangsu
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service