All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H5Cl3N2S
CAS Number:
Molecular Weight:
255.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
97%
form
solid
mp
206-211 °C (lit.)
functional group
amine
chloro
thiourea
SMILES string
NC(=S)Nc1c(Cl)cc(Cl)cc1Cl
InChI
1S/C7H5Cl3N2S/c8-3-1-4(9)6(5(10)2-3)12-7(11)13/h1-2H,(H3,11,12,13)
InChI key
ZUSZPZPGZMEXLA-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service