Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

558524

Sigma-Aldrich

2-Methylthiothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NS2
CAS Number:
Molecular Weight:
131.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.6080 (lit.)

bp

205-207 °C (lit.)

density

1.271 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

CSc1nccs1

InChI

1S/C4H5NS2/c1-6-4-5-2-3-7-4/h2-3H,1H3

InChI key

VQNOAXZUEKPSJC-UHFFFAOYSA-N

Application

2-Methylthiothiazole may be used for the preparation of 1-(2-methylthio-5-thiazolyl)ethanol.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

195.1 °F - closed cup

flash_point_c

90.6 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transmission of substituent effects in heterocyclic systems. Rates of solvolysis of substituted thiazolylethanol derivatives.
Noyce DS and Fike SA.
The Journal of Organic Chemistry, 38(19), 3318-3321 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service