Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

554391

Sigma-Aldrich

2-(Trifluoromethoxy)iodobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4IOCF3
CAS Number:
Molecular Weight:
288.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5060 (lit.)

bp

164-165 °C (lit.)

density

1.855 g/mL at 25 °C (lit.)

functional group

fluoro
iodo

SMILES string

FC(F)(F)Oc1ccccc1I

InChI

1S/C7H4F3IO/c8-7(9,10)12-6-4-2-1-3-5(6)11/h1-4H

InChI key

GYBMJVZOZTVDKS-UHFFFAOYSA-N

Application

2-(Trifluoromethoxy)iodobenzene [1-Iodo-2-(trifluoromethoxy)benzene] may be used to synthesize 2-(trifluoromethoxy)biphenylyl-2′-diazonium salts.[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

150.1 °F - closed cup

flash_point_c

65.6 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CF3 Oxonium Salts, O-(Trifluoromethyl) dibenzofuranium Salts: In Situ Synthesis, Properties, and Application as a Real CF3+ Species Reagent.
Umemoto T, et al.
The Journal of Organic Chemistry, 72(18), 6905-6917 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service