Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

552461

Sigma-Aldrich

2-Fluoro-5-iodobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(I)COCl
CAS Number:
Molecular Weight:
284.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

30-34 °C (lit.)

SMILES string

Fc1ccc(I)cc1C(Cl)=O

InChI

1S/C7H3ClFIO/c8-7(11)5-3-4(10)1-2-6(5)9/h1-3H

Inchi Key

CEMYZMDITJKYDA-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of biologically and pharmacologically active molecules
  • Microwave-assisted boron trichloride mediated coupling reactions

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service