Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

551910

Sigma-Aldrich

2-Hydroxy-5-methyl-3-nitropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay:
97%

assay

97%

mp

179 °C (dec.) (lit.)

functional group

nitro

SMILES string

Cc1cnc(O)c(c1)[N+]([O-])=O

InChI

1S/C6H6N2O3/c1-4-2-5(8(10)11)6(9)7-3-4/h2-3H,1H3,(H,7,9)

InChI key

QAINEQVHSHARMD-UHFFFAOYSA-N

Application

2-Hydroxy-5-methyl-3-nitropyridine may be used in the preparation of 2-chloro-5-methyl-3-nitropyridine via chlorination, using thionyl chloride. It may also be used is preparing proteasome inhibitors containing 5-methylpyridin-2(1H)-one moiety.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Chloro-5-methyl-3-nitropyridine.
Zhang DT and Huo LY.
Acta Crystallographica Section E, Structure Reports Online, 67(11), o3134-o3134 (2011)
Incorporation of the Constrained Peptidomimetic, 5-Methylpyridin-2-one into Peptide Vinyl Sulfones and Peptide Epoxy Ketones is Detrimental for Proteasome Inhibition.
Xin BT, et al.
European Journal of Organic Chemistry (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service