Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

550000

Sigma-Aldrich

3-Isopropoxypropylamine

≥98%

Synonym(s):

IPOPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHO(CH2)3NH2
CAS Number:
Molecular Weight:
117.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98%

refractive index

n20/D 1.4195 (lit.)

bp

78-79 °C/85 mmHg (lit.)

density

0.845 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

CC(C)OCCCN

InChI

1S/C6H15NO/c1-6(2)8-5-3-4-7/h6H,3-5,7H2,1-2H3

InChI key

VHYUNSUGCNKWSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Isopropoxypropylamine is an aliphatic primary amine.

Application

3-Isopropoxypropylamine may be used as a reactant in the preparation of:
  • 1-alcoxyalkyl-5-aryl-4-acyl-3-hydroxy-3-pyrrolin-2-ones[1]
  • 1-(p-sulfamyl)-phenyl-5-oxo-3-pyrrolidine carboxylic acid derivative[2]
  • ethyl 1,4-dihydro-1-(3-isopropoxypropyl)-4-oxo-3-quinolinecarboxylate[3]

Legal Information

Product of Arkema Inc.

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl 1, 4-dihydro-4-oxo-3-quinolinecarboxylates by a tandem addition-elimination-SNAr reaction.
Bunce RA, et al.
Journal of Heterocyclic Chemistry, 48(3), 620-625 (2011)
Synthesis and antibacterial activity of 1-alkoxyalkyl-5-aryl-4-acyl-3-hydroxy-3-pyrrolin-2-ones.
Gein VL, et al.
Pharmaceutical Chemistry Journal, 41(4), 208-210 (2007)
Itaconic acid derivatives of sulfanilamide.
Paytash PL, et al.
Journal of the American Chemical Society, 74(18), 4549-4552 (1952)
Jeffrey A Schneider et al.
Nature communications, 9(1), 4396-4396 (2018-10-26)
New chemical inhibitors of protein-protein interactions are needed to propel advances in molecular pharmacology. Peptoids are peptidomimetic oligomers with the capability to inhibit protein-protein interactions by mimicking protein secondary structure motifs. Here we report the in silico design of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service