544582
Zinc naphthenate
<8% (Zn by wt.)
Synonym(s):
Naphthalic acid zinc salt solution
About This Item
Recommended Products
concentration
≤50% in mineral spirits
<8% Zn (by wt.)
impurities
≤8% diethylene glycol monoethyl ether
refractive index
n20/D 1.4630 (lit.)
density
0.962 g/mL at 25 °C (lit.)
Physical form
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 3 - STOT RE 1
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
135.0 °F - closed cup
flash_point_c
57.2 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service