Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538744

Sigma-Aldrich

4-Iodophenetole

97%

Synonym(s):

4-Iodophenyl ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4OCH2CH3
CAS Number:
Molecular Weight:
248.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

25-28 °C (lit.)

functional group

iodo

SMILES string

CCOc1ccc(I)cc1

InChI

1S/C8H9IO/c1-2-10-8-5-3-7(9)4-6-8/h3-6H,2H2,1H3

InChI key

VSIIHWOJPSSIDI-UHFFFAOYSA-N

Application

4-Iodophenetole may be used as one of the components in the synthesis of bis(4-ethoxyphenyl)amine.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of 5-[bis (4-ethoxyphenyl) amino] thiophene-2-carbaldehyde
Tan, J-Y., Ming Kong, and J-Y. Wu
Acta Crystallographica Section E, Structure Reports Online, 70.9, o1075-o1076 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service