Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530271

Sigma-Aldrich

Methyl 3-amino-2-methylbenzoate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)CO2CH3
CAS Number:
Molecular Weight:
165.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.573 (lit.)

bp

300 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1cccc(N)c1C

InChI

1S/C9H11NO2/c1-6-7(9(11)12-2)4-3-5-8(6)10/h3-5H,10H2,1-2H3

InChI key

ZOOQFAUFPWXUMI-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service